missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Anti-Human IgG Monoclonal Antibody from Mouse

Product Code. 16399845 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
0.5 mg
Unit Size:
0.50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16399845 0.5 mg 0.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16399845 Supplier MP Biomedicals™ Supplier No. 08634811

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Anti-Human IgG Monoclonal Antibody from Mouse, in 10 mM Phosphate Buffer at pH 7.4, containing 0.9% NaCl and 0.02% NaN3.

  • Anti-Human IgG Monoclonal Antibody from Mouse, in 10 mM Phosphate Buffer at pH 7.4, containing 0.9 % NaCl and 0.02% NaN3
TRUSTED_SUSTAINABILITY

Specifications

Classification Monoclonal
Clone HIgG01
Formulation 10 mM Phosphate Buffer at pH 7.4, containing 0.9% NaCl and 0.02% NaN3
Host Species Mouse
Quantity 0.5 mg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Human
Content And Storage Store at -20°C for long term and 2 to 8°C for short term
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.