missing translation for 'onlineSavingsMsg'
Learn More

BD Biosciences™ Anti-Human Lineage Cocktail 2 (lin 2) (CD3, CD14, CD19, CD20, CD56) (RUO/GMP)

Product Code. 15806309 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15806309 50 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15806309 Supplier BD Biosciences™ Supplier No. 643397
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Monoclonal antibodies for detecting human antigens.

  • Clone NCAM16.2 (also known as NCAM 16) MφP9 (also known as MφP-9) SK7 (also known as Leu-4) SJ25C1 (also known as SJ25-C1) L27 (RUO (GMP))
  • Mouse IgG1 heavy chains and kappa light chains:
    • CD3, clone SK7
    • CD19, clone SJ25C1
    • CD20, clone L27
  • Mouse IgG2b heavy chains and kappa light chains:
    • CD14, clone MφP9
    • CD56, clone NCAM16.2
  • The Lineage Cocktail 2 (lin 2) contains antibody clones that, in combination, stain lymphocytes, monocytes, eosinophils, and neutrophils.
  • It includes CD3, CD14, CD19, CD20, and CD56. Peripheral blood dendritic cells (DCs), including those expressing CD16, can be distinguished from other leucocytes by their lack of staining with lin 2.
  • Contents:
  • Specificity: CD56 (NCAM-1), Clone:NCAM 16.2 (NCAM 16), Format: FITC, Isotype: IgG2bK, Entrez Gen ID: 4684
  • Specificity: CD14:, Clone: MφP9 (MφP-9), Format: FITC, Isotype: IgG2bK
  • Specificity: CD3, Clone: SK7(Leu-4), Format: FITC, Isotype: IgG1K, Entrez Gen ID: 916
  • Specificity: CD19, Clone: SJ25C1 (SJ25-C1), Format FITC, Isotype: IgG1K, Entrez Gen ID: 930
  • Specificity: CD20, Clone: L27, Format: FITC, Isotype: IgG1K, Entrez Gen ID: 931
  • Research applications include the following:
    • Dendritic cell (DC) subsets and basophils in peripheral blood.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD3, CD14, CD19, CD20, CD56
Applications Flow Cytometry
Clone NCAM16.2, MφP9, SK7, SJ25C1, L27
Concentration 40.6 μg/mL
Host Species Mouse
Quantity 50 Tests
Regulatory Status RUO
Includes Phosphate buffered saline with gelatin and 0.1% sodium azide.
Target Species Human
Content And Storage Store vials at 2°C–8°C. Conjugated forms should not be frozen. Protect from exposure to light. Each reagent is stable until the expiration date shown on the bottle label when stored as directed.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.