missing translation for 'onlineSavingsMsg'
Learn More

BD Biosciences™ Anti-Human Lineage Cocktail 3 (lin 3) (CD3, CD14, CD19, CD20) (RUO/GMP)

Product Code. 15816309
Change view
Click to view available options
Quantity:
50 Tests
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15816309 50 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15816309 Supplier BD Biosciences™ Supplier No. 643510
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Monoclonal antibodies for detecting human antigens.

  • Clone SK7 (also known as Leu-4) L27 SJ25C1 (also known as SJ25-C1) MφP9 (also known as MφP-9) (RUO (GMP))
  • Mouse IgG1 heavy chains and kappa light chains:
    • CD3, clone SK7
    • CD19, clone SJ25C1
    • CD20, clone L27
  • Mouse IgG2b heavy chains and kappa light chains:
    • CD14, clone MφP9
  • The Lineage Cocktail3 (lin 3) contains antibody clones that, in combination, stain lymphocytes, monocytes, eosinophils, and neutrophils.
  • It includes CD3, CD14, CD19, and CD20. Peripheral blood dendritic cells (DCs), including those expressing CD16, can be distinguished from other leucocytes by their lack of staining with lin 3.
  • Contents:
  • Specificity: CD14:, Clone: MφP9 (MφP-9), Format FITC, Isotype: IgG2bK
  • Specificity: CD3, Clone: SK7(Leu-4), Format: FITC, Isotype: IgG1K, Entrez Gen ID: 916
  • Specificity: CD19, Clone: SJ25C1 (SJ25-C1), Format: FITC, Isotype: IgG1K, Entrez Gen ID: 930
  • Specificity: CD20, Clone: L27, Format: FITC, Isotype: IgG1K, Entrez Gen ID: 931
  • Research applications include the following:
    • Dendritic cells (DC) in peripheral blood.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD3, CD20, CD19, CD14
Applications Flow Cytometry
Clone SK7, L27, SJ25C1, MφP9
Concentration 37.6 μg/mL
Host Species Mouse
Quantity 50 Tests
Regulatory Status RUO
Includes Phosphate buffered saline with gelatin and 0.1% sodium azide.
Target Species Human
Content And Storage Store vials at 2°C–8°C. Conjugated forms should not be frozen. Protect from exposure to light. Each reagent is stable until the expiration date shown on the bottle label when stored as directed.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.