Learn More
anti-IgG1, κ, Mouse, Alexa Fluor™ 488, Clone: MOPC-21, BD
Description
The MOPC-21 immunoglobulin is a mouse myeloma protein. The MOPC-21 immunoglobulin was selected as an isotype control following screening for low background on a variety of mouse and human tissues. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: MOPC-21
Isotype: IgG1 κ
Species Reactivity: Human
Flow Cytometry, Intracellular Staining, Isotype Control
Spécification
Spécification
| Applications | Flow Cytometry, Western Blot |
| Clone | MOPC-21 |
| Conjugate | Alexa Fluor 488 |
| Host Species | Mouse |
| Purification Method | Affinity Purified |
| Quantity | 100 Tests |
| Target Species | Human |
| Content And Storage | Undiluted at 4°C |
| Isotype | IgG1 κ |
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.