missing translation for 'onlineSavingsMsg'
Learn More

anti-IgG1 κ, Mouse , PE, Clone: MOPC-21, BD

Product Code. 15808778
Change view
Click to view available options
Quantity:
50 Tests
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15808778 50 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15808778 Supplier BD Biosciences Supplier No. 556650

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The MOPC-21 immunoglobulin is a mouse myeloma protein. The MOPC-21 immunoglobulin was selected as an isotype control following screening for low background on a variety of mouse and human tissues. This reagent has been used as isotype control in immunophenotypic studies with baboon, rhesus, or cynomolgus peripheral blood cells. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Mouse
Clone: MOPC-21
Isotype: IgG1, κ
Species Reactivity: Baboon/Rhesus/Cynomolgus

Flow Cytometry, Isotype Control

TRUSTED_SUSTAINABILITY

Specifications

Applications Flow Cytometry
Clone MOPC-21
Conjugate PE
Host Species Mouse
Purification Method Affinity Purified
Quantity 50 Tests
Target Species Baboon, Cynomolgus, Rhesus Monkey
Content And Storage Undiluted at 4°C
Isotype IgG1 κ
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.