missing translation for 'onlineSavingsMsg'
Learn More

anti-IgG1, PE, Clone: X40, BD

Product Code. 17899290
Change view
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
Each
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17899290 100 μg Pack of 100
15815848 50 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17899290 Supplier BD Biosciences Supplier No. 349043
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
The Mouse IgG1 antibody, clone X40, is derived from the hybridization of Sp2/0-Ag 14 mouse myeloma cells with spleen cells from BALB/c mice immunized with KLH. The antibodies react specifically with keyhole limpet hemocyanin (KLH), an antigen not expressed on human cells or human cell lines.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG1
Classification Monoclonal
Clone X40
Conjugate PE
Host Species Mouse
Quantity 100 μg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Isotype IgG1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.