missing translation for 'onlineSavingsMsg'
Learn More

anti-IgG2, κ, Clone: B81-3, BD

Product Code. 15819698
Change view
Click to view available options
Concentration:
0.5mg/mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Concentration Unit Size
15819698 0.5mg/mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15819698 Supplier BD Biosciences Supplier No. 559277
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The B81-3 antibody is specific for keyhole limpet hemocyanin. This antibody is routinely tested by LAL assay for endotoxin levels. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Blocking Assay, Co-Stimulation, Cytotoxicity Assay, Isotype Control

TRUSTED_SUSTAINABILITY

Specifications

Applications Blocking Assay, Functional Assay, Cytotoxicity Assay
Clone B81-3
Concentration 0.5mg/mL
Conjugate Unconjugated
Host Species Hamster
Purification Method Affinity Purified
Quantity 0.5 mg
Content And Storage Undiluted at 4°C
Isotype IgG2 κ
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.