missing translation for 'onlineSavingsMsg'
Learn More

anti-IgG2a, κ, Clone: G155-178, BD

Product Code. 15838278
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15838278 0.1 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15838278 Supplier BD Biosciences Supplier No. 555571
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

The G155-178 clone has an unknown specificity. Trinitrophenal (TNP), the immunogen, is a hapten not expressed on human, mouse, rat or non-human primate cells. In the absence of specific binding, this antibody may bind non-specifically to Fc receptors. The immunoglobulin from clone G155-178 was selected as an isotype control following screening for low background on a variety of mouse and human tissues. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Flow Cytometry, Isotype Control

TRUSTED_SUSTAINABILITY

Specifications

Applications Flow Cytometry
Clone G155-178
Concentration 0.5mg/mL
Conjugate Unconjugated
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.1 mg
Content And Storage Undiluted at 4°C
Isotype IgG2a κ
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.