missing translation for 'onlineSavingsMsg'
Learn More

anti-IgG2a κ , Clone: R35-95, BD

Product Code. 15819678
Change view
Click to view available options
Quantity:
0.25 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15819678 0.25 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15819678 Supplier BD Biosciences Supplier No. 559073
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

The R35-95 hybridoma was generated by hybridization of Y3 myeloma cells with spleen cells from LOU rats immunized with mouse immunoglobulins. The R35-95 hybridoma produces rat IgG2a, κ immunoglobulin that has no measurable reactivity with mouse immunoglobulins. The R35-95 immunoglobulin was selected as an isotype control following screening for low background binding on a variety of mouse and human tissues.

Immunocytochemistry, Immunohistochemistry (Formalin), Immunohistochemistry (Frozen), Immunohistochemistry (Zinc-Fixed), Isotype Control

TRUSTED_SUSTAINABILITY

Specifications

Applications Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Zinc-Fixed)
Clone R35-95
Concentration 0.5mg/mL
Conjugate Unconjugated
Host Species Rat
Purification Method Affinity Purified
Quantity 0.25 mg
Content And Storage Undiluted at 4°C
Isotype IgG2a κ
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.