Learn More
anti-IgG2a κ, Rat , Alexa Fluor™ 647, Clone: R35-95, BD
185.62 € - 371.24 €
Specifications
| Clone | R35-95 |
|---|---|
| Applications | Flow Cytometry, Western Blot |
| Conjugate | Alexa Fluor 647 |
| Host Species | Rat |
| Immunogen | Mouse Pooled Immunoglobulin |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15878988
|
BD Biosciences
557690 |
0.1 mg |
387.00 € 371.24 € / Each Save 15.76 € 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15869098
|
BD Biosciences
557906 |
100 Tests |
194.00 € 185.62 € / Each Save 8.38 € 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The R35-95 hybridoma was generated by hybridization of Y3 myeloma cells with spleen cells from LOU rats immunized with mouse immunoglobulins. The R35-95 hybridoma produces rat IgG2a, κ immunoglobulin that has no measurable reactivity with mouse immunoglobulins. The R35-95 immunoglobulin was selected as an isotype control following screening for low background binding on a variety of mouse and human tissues. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Rat
Clone: R35-95
Isotype: IgG2a, κImmunogen: Mouse Pooled Immunoglobulin
Flow Cytometry, Intracellular Staining, Isotype Control
Specifications
| R35-95 | |
| Alexa Fluor 647 | |
| Mouse Pooled Immunoglobulin | |
| Affinity Purified |
| Flow Cytometry, Western Blot | |
| Rat | |
| IgG2a κ | |
| Undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.