missing translation for 'onlineSavingsMsg'
Learn More

anti-IgG2b κ, Rat, FITC, Clone: A95-1, BD

Brand:  BD Biosciences 556923

Product Code. 15878798

  • ONLINE EXCLUSIVE PRICE
    235.00 €
    225.77 € / Each
    Save 9.23 €
    3% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The A95-1 antibody has unknown specificity. Trinitrophenal (TNP), the immunogen, is a hapten not expressed on human, mouse, rat, or non-human primate cells. The A95-1 immunoglobulin was selected as an isotype control following screening for low background on a variety of mouse and human tissues. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Rat
Clone: A95-1
Isotype: IgG2b, κ
Species Reactivity: Mouse
Immunogen: TNP-Keyhole Limpet Hemocyanin

Flow Cytometry, Isotype Control

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Flow Cytometry
0.5mg/mL
Rat
Affinity Purified
Mouse
IgG2b κ
A95-1
FITC
TNP-keyhole limpet hemocyanin
0.1 mg
Undiluted at 4°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.