missing translation for 'onlineSavingsMsg'
Learn More

anti-IgM, Brilliant Violet 786, Clone: R6-60.2, BD

Product Code. 15843299
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15843299 50 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15843299 Supplier BD Biosciences Supplier No. 564028

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Host Species: Rat
Clone: R6-60.2
Isotype: IgG2a, κ
Species Reactivity: Ms
Immunogen: Pooled Mouse Ig

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen IgM
Classification Monoclonal
Clone R6-60.2
Concentration 0.2mg/mL
Conjugate Brilliant Violet 786
Host Species Rat
Purification Method Affinity Purified
Quantity 50 μg
Primary or Secondary Secondary
Target Species Mouse
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.