missing translation for 'onlineSavingsMsg'
Learn More

anti-IgM, κ, Clone: G155-228, BD

Product Code. 15808288
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15808288 0.1 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15808288 Supplier BD Biosciences Supplier No. 555581
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

The G155-228 antibody is specific for the hapten trinitrophenol (TNP), a hapten not expressed on non-human primate cells or non-human primate cell lines. In the absence of specific binding, this immunoglobulin may bind non-specifically to Fc receptors. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Applications Flow Cytometry
Clone G155-228
Concentration 0.5mg/mL
Conjugate Unconjugated
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.1 mg
Content And Storage Undiluted at 4°C
Isotype IgM κ
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.