Learn More
IRE1 alpha (p Ser724) Rabbit anti-Human, Mouse, Rat, Porcine, Drosophila, Goat, Mammalia, Monkey, Primate, Rabbit, Golden Syrian Hamster, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 252 publications
Brand: Novus Biologicals NB100-2323
Description
IRE1 alpha Polyclonal specifically detects IRE1 alpha in Human, Mouse, Rat, Porcine, Drosophila, Goat, Mammal, Monkey, Primate, Rabbit, Golden Syrian Hamster samples. It is validated for Western Blot, Chromatin Immunoprecipitation, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, In vitro assay, Knockdown Validated.Specifications
| IRE1 alpha (p Ser724) | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| EC 2.7.11, endoplasmic reticulum to nucleus signaling 1, Endoplasmic reticulum-to-nucleus signaling 1, ER to nucleus signalling 1, FLJ30999, hIRE1p, inositol-requiring 1, Inositol-requiring protein 1, IRE 1a, ire1 alpha p Ser724, ire1 alpha phosphorylation, IRE1a, ire1-alpha, IRE1MGC163279, IRE1P, MGC163277, phospho ire1 alpha, phospho-ire1 alpha, phospho-ire1a, p-ire1 alpha, protein kinase/endoribonuclease, serine/threonine-protein kinase/endoribonuclease IRE1 | |
| Rabbit | |
| 110 kDa | |
| 0.1 mL | |
| Primary | |
| NB100-2323 IRE1 alpha [p Ser724] Antibody detects IRE-1 alpha when phosphorylated at Ser724 residue. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, ChIP assay, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| 1.0 mg/mL | |
| Western Blot 1:1000, Chromatin Immunoprecipitation, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500, Immunohistochem | |
| O75460 | |
| ERN1 | |
| This IRE1 alpha [p Ser724] antibody was raised against a synthetic peptide surrounding the phosphorylated serine 724 of the human IRE1 alpha protein. [Swiss-Prot #O75460] | |
| Affinity Purified | |
| RUO | |
| 2081 | |
| Human, Mouse, Rat, Porcine, Drosophila, Goat, Mammalia, Monkey, Primate, Rabbit, Golden Syrian Hamster | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
For Research Use Only