Learn More
anti-Lymphotoxin beta/TNFSF3, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-14207
Encompass
Product Code. 15705436
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Lymphotoxin beta/TNFSF3 Polyclonal specifically detects Lymphotoxin beta/TNFSF3 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.