missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Anti-miR™ hsa-let-7c miRNA Inhibitor Positive Control

Product Code. 10554065
Change view
Click to view available options
Quantity:
5 nmol
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10554065 5 nmol Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10554065 Supplier Invitrogen™ Supplier No. 4392431
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Anti-miR™ hsa-let-7c miRNA Inhibitor Positive Control

Invitrogen™ Anti-miR™ miRNA Inhibitors are chemically modified, single-stranded nucleic acids designed to specifically bind to and inhibit endogenous microRNA (miRNA) molecules. These ready-to-use inhibitors can be introduced into cells using transfection or electroporation parameters similar to those used for siRNAs. Anti-miR™ let-7c miRNA Inhibitor Positive Control provides a convenient, validated positive control for experiments using Anti-miR miRNA Inhibitors. Endogenous let-7 miRNA negatively regulates HMGA2, a ubiquitously expressed, nonhistone chromatin protein that modulates gene expression through changes in chromatin architecture. When transfected into human and mouse cells, Anti-miR hsa-let-7c miRNA Inhibitor blocks endogenous let-7c miRNA, resulting in increased levels of HMGA2 mRNA.

Accessory Products:
Also available are Anti-miR™ miRNA Inhibitors (SKU #AM17000 and #AM17001). To select your miRNA sequence and order, please visit the GeneAssist™ miRNA Workflow Builder. Anti-miR™ miRNA Inhibitor Negative Control #1 (SKU #AM17010) is a random sequence that has been validated to produce no identifiable effects on known miRNA function. Cy™3 and FAM™ dye-labeled Anti-miR Negative Controls (SKU #AM17011 and #AM17012, respectively) are also available. The Pre-miR™ miRNA Starter Kit (SKU #AM1540) can be used to optimize transfection of Pre-miR™ miRNA Precursors.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Anti-miR™ miRNA is provided dried down in a single tube, along with Nuclease-free Water for resuspension, and should be stored at –20°C. It is guaranteed for 1 year when stored dried at –20°C.
Control Type Positive Control
RNAi Type miRNA Inhibitor
For Use With (Application) miRNA & Non-Coding RNA Analysis
Label or Dye Unconjugated
Product Line Ambion, Anti-miR
Product Type miRNA Inhibitor
Quantity 5 nmol
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.