missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-CD19, Clone: CB19, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 9 publications
Brand: Novus Biologicals NBP2-25196
This item is not returnable.
View return policy
Description
CD19 Monoclonal specifically detects CD19 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, In vitro assay, CyTOF-ready.Specifications
| CD19 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry 0.2 ug/10^6 cells, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:25-1:100, In vitro assay, CyTOF-ready | |
| P15391 | |
| CD19 | |
| Intact normal human B cell cells were used as immunogen to generate the CB19 clone. | |
| 0.1 mg | |
| Primary | |
| This product reacts with Human. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, In vitro Assay | |
| CB19 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| B4, B-lymphocyte antigen CD19, B-lymphocyte surface antigen B4, CD19 antigen, CD19 molecule, CVID3, Differentiation antigen CD19, MGC12802, T-cell surface antigen Leu-12 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 930 | |
| Human, Mouse | |
| Purified |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?
For Research Use Only