missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-GSK-3 beta, Clone: 3D10, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 6 publications
Brand: Novus Biologicals NBP1-47470SS
This item is not returnable.
View return policy
Description
GSK-3 beta Monoclonal specifically detects GSK-3 beta in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Simple Western, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| GSK-3 beta | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:500-1:2000, Simple Western 1:50, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| P49841 | |
| GSK3B | |
| Purified recombinant fragment of human GSK3 beta expressed in E. coli. [UniProt# P49841] | |
| Ammonium sulfate precipitation | |
| RUO | |
| 2932 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 3D10 | |
| Unconjugated | |
| PBS with 0.03% Sodium Azide | |
| EC 2.7.11, EC 2.7.11.26, glycogen synthase kinase 3 beta, glycogen synthase kinase-3 beta, GSK-3 beta, GSK3beta isoform | |
| Mouse | |
| 46 kDa | |
| 0.025 mL | |
| Primary | |
| Human, Mouse, Rat, Primate | |
| Precipitation |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
For Research Use Only