missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-Histone H3 (Trimethyl Lys9), Clone: 6F12-H4, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 19 publications
Brand: Novus Biologicals NBP1-30141SS
This item is not returnable.
View return policy
Description
Histone H3 Monoclonal specifically detects Histone H3 in Human, Mouse, Rat, Porcine, C. elegans, Drosophila, Invertebrate, Mammal, Yeast samples. It is validated for Western Blot, Chromatin Immunoprecipitation, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| Histone H3 (Trimethyl Lys9) | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:2000, Chromatin Immunoprecipitation 1:10-1:500, ELISA 1:100-1:2000, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:200, Immunohistochemistry-Paraffin 1:1000 | |
| P84243, P84244 | |
| H3C14 | |
| This Histone H3 [Trimethyl Lys9] antibody (6F12-H4) was raised against a synthetic peptide made to an N-terminal region of Histone H3 (between amino acids 1-50). [UniProt# P84243] | |
| Protein G purified | |
| RUO | |
| 126961 | |
| Human, Mouse, Rat, Porcine, C. elegans, Drosophila, Invertebrate, Mammalia, Yeast | |
| Purified |
| Western Blot, ChIP assay, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 6F12-H4 | |
| Unconjugated | |
| Tris-Glycine and 0.15M NaCl with 0.05% Sodium Azide | |
| H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, H3K9Me3, HIST2H3C, histone H3.3, MGC87782, MGC87783 | |
| Mouse | |
| 15 kDa | |
| 0.025 mL | |
| Primary | |
| All mammals including human, mouse and rat, Drosophila, C. elegans, yeast. Invertebrate/Blattella germanica (German cockroach) reactivity reported in scientific literature (PMID: 23872316). | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
For Research Use Only