Learn More
Phospho-RelA/NFkB p65 (S536) Rabbit anti-Human, Clone: 1091B, R&D Systems™
GREENER_CHOICE
Rabbit Monoclonal Antibody has been used in 3 publications
Brand:  R&D Systems MAB72261
Encompass
Product Code. 15709134
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
RelA/NFkB p65 Monoclonal specifically detects RelA/NFkB p65 in Human samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Contains <0.1% Sodium Azide, which is not hazardous at this concentration according to GHS classifications. Refer to SDS for additional information and handling instructions.