missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Anti-SARS-CoV-2 Antibody IgM Titer Serologic Assay kit (S1 protein)
Click to view available options
Quantity:
96 Tests
Unit Size:
96 tests
Description
The newly identified Severe Acute Respiratory Syndrome Coronavirus 2 (SARS-CoV-2) has posed a serious threat to human health. A rapid and effective assay kit detecting the levels of anti-SARS-CoV-2 in human serum can facilitate research on characterization of antibodies produced in response to SARS-CoV-2 infection.
Specifications
Specifications
| Product Type | Antibody titer assay kits |
| Kit Contents | This assay kit employs a standard capture-ELISA format, providing a rapid detection of anti-SARS-CoV-2 antibody, IgM in serum by SARS-CoV-2 S1 protein. The kit consists of HPR-S1 protein, an Anti-SARS-CoV-2 Antibody (Control, IgM), an anti-Human IgM anto |
| Quantity | 96 Tests |
| Shipping Condition | RT |
| Storage Requirements | -20°C, aviod light |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction