Learn More
Sartorius Anti-Static Devices for Balances
GREENER_CHOICE
Product Code. 10465765 Shop All Sartorius Products
Change view
Click to view available options
Type:
Anti-static Pen
Unit Size:
Each
Product Code. Type unitSize
10465765 Anti-static Pen Each
1 options
Product Code. 10465765
Brand: Sartorius YSTP01
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Control charges on samples
  • Anti-static pen—direct the pen at item to be neutralized
  • Ionizing blower—point blower toward sample prior to weighing
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) CPA Balance, Extended Balance, SE and ME Ultramicro Balance
Type Anti-static Pen