Learn More
Buerkle™ Anti-Static Set for Decanting Flammables
Product Code. 10057710 Shop All Bürkle Products
Change view
Click to view available options
Includes:
(3) Copper Cable; Clamp
Unit Size:
Each
Product Code. Includes Unit Size
10057710 (3) Copper Cable; Clamp Each
1 options
Product Code. 10057710 Supplier Buerkle™ Supplier No. 56021000
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Buerkle™ Anti-Static Set for Decanting Flamables ensures easy and reliable earthing.
  • When decanting flammable liquids without the machines and containers involved in the process being sufficiently earthed, electrostatic build-up can occur, which for many substances can result in fire or an explosion.
  • Anti-static set ensures easy and reliable earthing
  • The Anti-static set consists of three colour-coded connecting copper cables with clamps in a handy plastic case
TRUSTED_SUSTAINABILITY
Specifications
Description For easy and reliable earthing
Includes (3) Copper Cable; Clamp
For Use With Electric Conductive Pumps