missing translation for 'onlineSavingsMsg'
Learn More

BD Pharmingen™ PE Streptavidin

Product Code. 15837558
Change view
Click to view available options
Quantity:
0.5 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15837558 0.5 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15837558 Supplier BD Supplier No. 554061

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

SAv-PE is a useful second-step reagent for the indirect immunofluorescent staining of cells in combination with biotinylated primary antibodies for flow cytometric analysis.

  • SAv-PE is a useful second-step reagent for the indirect immunofluorescent staining of cells in combination with biotinylated primary antibodies for flow cytometric analysis.
  • Excitation at 488-nm light leads to a fluorescence emission maximum of ∼575 nm.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.5 mg/mL
Conjugate PE (Phycoerythrin)
Quantity 0.5 mg
Regulatory Status For research use only (RUO)
Content And Storage Store undiluted at 4°C. Protect from prolonged exposure to light. Do not freeze.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.