Learn More
anti-TWEAK/TNFSF12, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Brand:  Novus Biologicals NBP1-76695
Encompass
Product Code. 15704746
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
TWEAK/TNFSF12 Polyclonal specifically detects TWEAK/TNFSF12 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.