Learn More
anti-Tyrosine Hydroxylase (p Ser40), Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 6 publications
Brand:  Novus Biologicals NB300-173
Encompass
Product Code. 15785237
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Tyrosine Hydroxylase Polyclonal specifically detects Tyrosine Hydroxylase in Mouse, Rat, Mammal samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.