missing translation for 'onlineSavingsMsg'
Learn More

Antibiotic A21 (Technical Grade), TRC

Product Code. 30348106
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30348106 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30348106 Supplier Toronto Research Chemicals Supplier No. TRCA69744010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Antibiotic A21 (Technical Grade)
CAS 1383659-59-3
Molecular Formula C54H82N4O18
Quantity 10 mg
Synonym Amphotericin A21 Molecule, L-Histidine, Methyl Ester, Amide With Amphotericin B
InChI Formula InChI=1S/C54H82N4O18/c1-31-18-16-14-12-10-8-6-7-9-11-13-15-17-19-39(75-53-50(68)47(55)49(67)34(4)74-53)26-44-46(51(69)58-40(52(70)72-5)22-35-29-56-30-57-35)43(64)28-54(71,76-44)27-38(61)24-42(63)41(62)21-20-36(59)23-37(60)25-45(65)73-33(3)32(2)48(31)66/h6-19,29-34,36-44,46-50,53,59-64,66-68,71H,20-28,55H2,1-5H3,(H,56,57)(H,58,69)/b7-6+,10-8+,11-9+,14-12+,15-13+,18-16+,19-17+/t31-,32-,33-,34+,36+,37+,38-,39-,40-,41+,42+,43-,44-,46+,47-,48+,49+,50-,53-,54+/m0/s1
SMILES COC(=O)[C@H](Cc1cnc[nH]1)NC(=O)[C@@H]2[C@@H](O)C[C@@]3(O)C[C@@H](O)C[C@@H](O)[C@H](O)CC[C@@H](O)C[C@@H](O)CC(=O)O[C@@H](C)[C@H](C)[C@H](O)[C@@H](C)\C=C\C=C\C=C\C=C\C=C\C=C\C=C\[C@@H](C[C@@H]2O3)O[C@@H]4O[C@H](C)[C@@H](O)[C@H](N)[C@@H]4O
IUPAC Name methyl (2S)-2-[[(1S,3R,4E,6E,8E,10E,12E,14E,16E,18S,19R,20R,21S,25R,27R,30R,31R,33S,35R,37S,38R)-3-[(2R,3S,4S,5S,6R)-4-amino-3,5-dihydroxy-6-methyloxan-2-yl]oxy-19,25,27,30,31,33,35,37-octahydroxy-18,20,21-trimethyl-23-oxo-22,39-dioxabicyclo[33.3.1]nonatriaconta-4,6,8,10,12,14,16-heptaene-38-carbonyl]amino]-3-(1H-imidazol-5-yl)propanoate
Molecular Weight (g/mol) 1075.24
Formula Weight 1074.5624
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.