missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Antibody Diluent, Normal, 125mL

Product Code. 11444346 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
125 mL
Unit Size:
125mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11444346 125 mL 125mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11444346 Supplier MP Biomedicals™ Supplier No. 08980641
  • ONLINE EXCLUSIVE PRICE
    165.00 €
    156.45 € / 125mL
    Save 8.55 €
    5% Off
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ready-to-use sterile phosphate buffered saline

Specifications

Chemical Name or Material Normal Antibody Diluent
For Use With (Application) Used as diluent, Immunoassays
Quantity 125 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.