Learn More
Tork Antimicrobial Foam Soap Refill S4
Product Code. 16679921 Shop All Tork Products
Change view
Click to view available options
Color:
White
Unit Size:
Pack of 6
Product Code. Color Unit Size
16679921 White Pack of 6
1 options
Product Code. 16679921 Supplier Tork Supplier No. 520800
Support a Safer, More Efficient Laboratory
Use promo code 28026
View all promotions
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Ensure effective use against a variety of problematic organisms.
  • Ensure effective use against a variety of problematic organisms - our antimicrobial soap refill conforms with EN 1499.
  • Reduce the risk of cross-contamination - our sealed refills come with a single-use pump for improved hygiene.
  • Intuitive refilling in less than 10 seconds means effortless maintenance for you and your staff.
  • Lower your eco-footprint and reduce waste with our recyclable bottle that collapses as the soap is used.
TRUSTED_SUSTAINABILITY
Specifications
Color White
Form Foam
Certifications/Compliance EN1499
Certifications/Compliance (Europe) EN1499
Disposable Limited-use
Packaging Plastic Bottle
Quantity 6/Pk.
Volume (Metric) 1 L