missing translation for 'onlineSavingsMsg'
Learn More

Antimony(III) chloride, 99.9% (metals basis)

Product Code. 11384288
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11384288 100 g 100g
11374288 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11384288 Supplier Thermo Scientific Alfa Aesar Supplier No. 041537.22
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10025-91-9 | Cl3Sb | 228.11 g/mol

Antimony(III) chloride is engaged to detect certain compounds, such as cholesterols. It is a complexing agent used in the colorimetric Carr-Price test for detecting carotenoids including vitamin A. It is employed as a mordant and a fabric flame retardant. It is used as a catalyst for chlorinationn and polymerization reactions. It is used to coat the steel to make bronze, used as a cutting material of antimony salt and as a preservative in medicine. It is used as an analytical reagent for chloral and aromatics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-91-9
Molecular Formula Cl3Sb
Molecular Weight (g/mol) 228.11
MDL Number MFCD00011212
InChI Key FAPDDOBMIUGHIN-UHFFFAOYSA-K
Synonym antimony trichloride, trichlorostibine, antimony chloride, antimony iii chloride, stibine, trichloro, antimontrichlorid, antimonous chloride, butter of antimony, antimony butter, caustic antimony
PubChem CID 24814
ChEBI CHEBI:74856
IUPAC Name trichlorostibane
SMILES [Cl-].[Cl-].[Cl-].[Sb+3]

Specifications

Boiling Point 223.5°C
Melting Point 73.4°C
Color White
Physical Form Crystals
Quantity 100 g
Odor Pungent
Sensitivity Moisture sensitive
Assay Percent Range 99.99% (Metals basis)
Density 3.14
Solubility Information Soluble in water,hydrochloric acid,alcohol,benzene,carbon disulfide,dioxane,chloroform,ether,acetone and carbon tetrachloride.
Formula Weight 228.11
Chemical Name or Material Antimony(III) chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.