missing translation for 'onlineSavingsMsg'
Learn More

Antimony(III) chloride, 99+%

Product Code. 11423944
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
Product Code. Quantity unitSize
11423944 100 g 100g
11433944 500 g 500g
11443944 2500 g 2500g
3 options
This item is not returnable. View return policy

Product Code. 11423944

Brand: Thermo Scientific Alfa Aesar A11944.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10025-91-9 | Cl3Sb | 228.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Antimony(III) chloride is engaged to detect certain compounds, such as cholesterols. It is a complexing agent used in the colorimetric Carr-Price test for detecting carotenoids including vitamin A. It is employed as a mordant and a fabric flame retardant. It is used as a catalyst for chlorinationn and polymerization reactions. It is used to coat the steel to make bronze, used as a cutting material of antimony salt and as a preservative in medicine. It is used as an analytical reagent for chloral and aromatics.

Solubility
Soluble in water, hydrochloric acid, alcohol, benzene, carbon disulfide, dioxane, chloroform, ether, acetone and carbon tetrachloride. Insoluble in organic bases.

Notes
Moisture sensitive and hygroscopic in nature. It is corrosive to metals and tissue. Incompatible with strong oxidizing agents, strong acids and strong bases. Decomposes in water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-91-9
Molecular Formula Cl3Sb
Molecular Weight (g/mol) 228.11
MDL Number MFCD00011212
InChI Key FAPDDOBMIUGHIN-UHFFFAOYSA-K
Synonym antimony trichloride, trichlorostibine, antimony chloride, antimony iii chloride, stibine, trichloro, antimontrichlorid, antimonous chloride, butter of antimony, antimony butter, caustic antimony
PubChem CID 24814
ChEBI CHEBI:74856
SMILES [Cl-].[Cl-].[Cl-].[Sb+3]

Specifications

Boiling Point 223.5°C
Melting Point 73.4°C
Quantity 100 g
UN Number UN1733
Merck Index 14,707
Solubility Information Soluble in water,hydrochloric acid,alcohol,benzene,carbon disulfide,dioxane,chloroform,ether,acetone and carbon tetrachloride. Insoluble in organic bases.
IUPAC Name trichlorostibane
Formula Weight 228.11
Percent Purity ≥99%
Odor Pungent
Sensitivity Moisture sensitive
Density 3.14
Chemical Name or Material Antimony (III) chloride
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Antimony(III) chloride
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Antimony trichloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.