missing translation for 'onlineSavingsMsg'
Learn More

Antimony(III Chloride, ReagentPlus™, 99%, Honeywell™

Product Code. 15682000 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Glass bottle
Unit Size:
500g
Product Code. Quantity Packaging unitSize
15682000 500 g Glass bottle 500g
1 options
This item is not returnable. View return policy

Product Code. 15682000

Brand: Honeywell Research Chemicals 215783500G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Boiling Point 283°C
Melting Point 73.4°C
CAS 10025-91-9
Color White
Physical Form Crystalline, Powder
Quantity 500 g
Molecular Formula Cl3Sb
Linear Formula SbCl3
MDL Number MFCD00011212
UN Number UN1733
Synonym antimony trichloride, trichlorostibine, antimony chloride, antimony iii chloride, stibine, trichloro, antimontrichlorid, antimonous chloride, butter of antimony, antimony butter, caustic antimony
InChI Key FAPDDOBMIUGHIN-UHFFFAOYSA-K
SMILES [Cl-].[Cl-].[Cl-].[Sb+3]
IUPAC Name antimony(3+) trichloride
Molecular Weight (g/mol) 228.11
PubChem CID 24814
ChEBI CHEBI:74856
Formula Weight 228.12g/mol
Percent Purity 99%
Grade ReagentPlus
Packaging Glass bottle
Vapor Pressure 1mmHg (49°C)
Density Vapor Density: 7.9 (vs air)
Chemical Name or Material Antimony(Iii) Chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.