missing translation for 'onlineSavingsMsg'
Learn More

Antimony(III) iodide, ultra dry, 99.998% (metals basis)

Product Code. 11495040
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11495040 1 g 1g
11363837 5 g 5g
11373837 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11495040 Supplier Thermo Scientific Alfa Aesar Supplier No. 035676.03
 Restricted Item
Estimated Shipment: 12-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7790-44-5 | I3Sb | 502.47 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-44-5
Molecular Formula I3Sb
Molecular Weight (g/mol) 502.47
MDL Number MFCD00016322
InChI Key KWQLUUQBTAXYCB-UHFFFAOYSA-K
Synonym antimony iii iodide, stibine, triiodo, antimony triiodide, antimon iii iodid, antimony iodide sbi3, antimony iodide, antimony iii iodide, ultra dry, antimony iii triiodide, hsdb 437, triiodostibine #
PubChem CID 24630
SMILES [Sb+3].[I-].[I-].[I-]

Tekniske data

Boiling Point 420°C
Melting Point 168°C
Physical Form Beads
Quantity 1 g
Odor Acrid
Packaging Ampule
Sensitivity Moisture sensitive
UN Number UN3260
Merck Index 14,710
Solubility Information Decomposed by water to SbOI.
IUPAC Name triiodostibane
Formula Weight 502.46
Percent Purity 99.998%
Assay (metals basis)
Chemical Name or Material Antimony(III) iodide, Ultra dry
Vis mere Vis mindre

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.