missing translation for 'onlineSavingsMsg'
Learn More

Antimony(III) oxide, 99.6% min

Product Code. 11387317
Change view
Click to view available options
Quantity:
50 g
500 g
2 kg
Unit Size:
2kg
500g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11387317 500 g 500g
11377317 50 g 50g
11397317 2 kg 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11387317 Supplier Thermo Scientific Alfa Aesar Supplier No. 011579.36
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1309-64-4 | O3Sb2 | 291.52 g/mol

It finds applications in flame retardants, catalysts, batteries, and alloys. It acts as an opacifying agent in glasses, ceramics and enamels. It is used as a catalyst in the manufacture of PET plastic (polyethylene terephthalate), and the vulcanization of rubber.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1309-64-4
Molecular Formula O3Sb2
Molecular Weight (g/mol) 291.52
MDL Number MFCD00011214
InChI Key GHPGOEFPKIHBNM-UHFFFAOYSA-N
Synonym Antimony trioxide
IUPAC Name diantimony(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Sb+3].[Sb+3]

Specifications

Boiling Point 1550°C (sublimation)
Melting Point 656°C
Color White
Physical Form Powder
Quantity 500 g
Assay Percent Range 99.6% min (Metal basis)
Solubility Information Poor solubility in water. Slowly dissolves in warm HCl or warm KOH solutions.
Formula Weight 291.5
Odor Odorless
Appearance White powder
Density 5.67
Chemical Name or Material Antimony(III) oxide
Show More Show Less
compliance-icons
Product Identifier
  • Antimony(III) oxide
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Antimony trioxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.