missing translation for 'onlineSavingsMsg'
Learn More

Antimony(III) oxide, 99.9% (metals basis)

Product Code. 11343398
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
Product Code. Quantity unitSize
11343398 50 g 50g
11333398 250 g 250g
2 options
This item is not returnable. View return policy

Product Code. 11343398

Brand: Thermo Scientific Alfa Aesar 036314.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1309-64-4 | O3Sb2 | 291.52 g/mol

Flame retardant for wide range of plastics, rubbers, paper and textiles . Catalyst in PET production. Activator in glass industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1309-64-4
Molecular Formula O3Sb2
Molecular Weight (g/mol) 291.52
MDL Number MFCD00011214
InChI Key GHPGOEFPKIHBNM-UHFFFAOYSA-N
IUPAC Name diantimony(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Sb+3].[Sb+3]

Specifications

Boiling Point 1550°C (sublimation)
Melting Point 656°C
Color White
Physical Form Sintered Lump
Quantity 50 g
Assay Percent Range 99.99% (Metals basis)
Solubility Information Very-slightly soluble in cold water. Slightly soluble in hot water,dilute sulfuric acid,and nitric acid. Soluble in solutions of alkali hydroxide and sulfides,hydrochloric acid,tartaric acid,and acetic acid
Formula Weight 291.5
Odor Odorless
Density 5.67 g/cm3
Chemical Name or Material Antimony(III) oxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.