missing translation for 'onlineSavingsMsg'
Learn More

Antimony(III) Oxide, TRC

Product Code. 30338078
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30338078 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30338078 Supplier Toronto Research Chemicals Supplier No. TRCA6975305G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Antimony Trioxide
CAS 1309-64-4
Quantity 5 g
Molecular Formula Sb2O3
Synonym Antimony oxide (Sb2O3), 100A, A 120, A 120 (corrosion inhibitor), ACC-BS, AN 800, AO 112, AO 3, AO 5, AP 50, AP 50 (metal oxide), AT 3, AT 3B, AT 3CN, Antimonious oxide, Antimony Bloom 100A, Antimony Bloom 500A, Antimony oxide, Antimony oxide (O3Sb2), Antimony oxide (SbO1.5), Antimony sesquioxide, Antimony trioxide, Antimony trioxide (Sb2O3), Antimony white, Antimony(3+) oxide, Antis N, Antis W, Antox, Atox B, Atox F, Atox R, Atox S, Bluestar RG, Bluestar Z, C.I. 77052, C.I. Pigment White 11, Chemetron Fire Shield, Dechlorane A-O, Diantimony trioxide, Exitelite, F 45, F 45 (oxide), FCP 100, FCP-AT 3, FCP-AT 3CN, FSPO 405, Fire Cut AT 3, Fire Cut AT 3CN, FireShield H, FireShield LS-FR, Fireshield FSPO 405, Flame Cut 610, Flame Cut 610R, Flameguard VF 59, Flowers of antimony, Garoflam SB 96DINP, HM 203P, HT 105, KN, KN (oxide), LO 112, LS-FR, LSB 80, MIC 3, MSA, MSA (flame retardant), MSF, Microfine AO 3, Microfine AO 5, NT 3H, Nyacol A 1510LP, Nyacol A 1530, Octoguard FR 10, P 3, PATOX-MF, Patox A, Patox C, Patox CF, Patox H, Patox HS, Patox K, Patox L, Patox M, Patox MK, Patox MZ, Patox P, Patox S, Patox SUF, Patox U, Performax 401, Poliflam HT 3, Polysafe 100T, Polysafe 60, Pyroguard AN 700, Pyroguard AN 800, Pyroguard AN 800T, Pyroguard AN 900, RAC 1, S 05N, SB 203, SBO-G, SHLB 80, Sanka Anchimonzol C, Sb-S, Selectipur 7835, Stibiox MS, Stibital, Stox W 60, TMS, TMS (flame retardant), TT 88, Thermoguard B, Thermoguard L, Thermoguard S, Timonox, Timonox RT, Timonox Red Star, Timonox Red Star TMS, Timonox White Star, Triox Blanc, Trutin 40, UF, UF (oxide), Ultrafine II, White Star, White Star N
InChI Formula InChI=1S/3O.2Sb
SMILES O=[Sb]O[Sb]=O
IUPAC Name oxo(oxostibanyloxy)stibane
Molecular Weight (g/mol) 291.52
Formula Weight 289.7924
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.