missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Antimony(III) Oxide, TRC
Tekniske data
Tekniske data
| Chemical Name or Material | Antimony Trioxide |
| CAS | 1309-64-4 |
| Quantity | 50 g |
| Molecular Formula | Sb2O3 |
| Synonym | Antimony oxide (Sb2O3), 100A, A 120, A 120 (corrosion inhibitor), ACC-BS, AN 800, AO 112, AO 3, AO 5, AP 50, AP 50 (metal oxide), AT 3, AT 3B, AT 3CN, Antimonious oxide, Antimony Bloom 100A, Antimony Bloom 500A, Antimony oxide, Antimony oxide (O3Sb2), Antimony oxide (SbO1.5), Antimony sesquioxide, Antimony trioxide, Antimony trioxide (Sb2O3), Antimony white, Antimony(3+) oxide, Antis N, Antis W, Antox, Atox B, Atox F, Atox R, Atox S, Bluestar RG, Bluestar Z, C.I. 77052, C.I. Pigment White 11, Chemetron Fire Shield, Dechlorane A-O, Diantimony trioxide, Exitelite, F 45, F 45 (oxide), FCP 100, FCP-AT 3, FCP-AT 3CN, FSPO 405, Fire Cut AT 3, Fire Cut AT 3CN, FireShield H, FireShield LS-FR, Fireshield FSPO 405, Flame Cut 610, Flame Cut 610R, Flameguard VF 59, Flowers of antimony, Garoflam SB 96DINP, HM 203P, HT 105, KN, KN (oxide), LO 112, LS-FR, LSB 80, MIC 3, MSA, MSA (flame retardant), MSF, Microfine AO 3, Microfine AO 5, NT 3H, Nyacol A 1510LP, Nyacol A 1530, Octoguard FR 10, P 3, PATOX-MF, Patox A, Patox C, Patox CF, Patox H, Patox HS, Patox K, Patox L, Patox M, Patox MK, Patox MZ, Patox P, Patox S, Patox SUF, Patox U, Performax 401, Poliflam HT 3, Polysafe 100T, Polysafe 60, Pyroguard AN 700, Pyroguard AN 800, Pyroguard AN 800T, Pyroguard AN 900, RAC 1, S 05N, SB 203, SBO-G, SHLB 80, Sanka Anchimonzol C, Sb-S, Selectipur 7835, Stibiox MS, Stibital, Stox W 60, TMS, TMS (flame retardant), TT 88, Thermoguard B, Thermoguard L, Thermoguard S, Timonox, Timonox RT, Timonox Red Star, Timonox Red Star TMS, Timonox White Star, Triox Blanc, Trutin 40, UF, UF (oxide), Ultrafine II, White Star, White Star N |
| InChI Formula | InChI=1S/3O.2Sb |
| SMILES | O=[Sb]O[Sb]=O |
| IUPAC Name | oxo(oxostibanyloxy)stibane |
| Molecular Weight (g/mol) | 291.52 |
| Formula Weight | 289.7924 |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?