missing translation for 'onlineSavingsMsg'
Learn More

Antimony potassium tartrate hydrate, 98%

Product Code. 11454243
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11454243 2500 g 2500g
11434243 100 g 100g
11444243 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11454243 Supplier Thermo Scientific Alfa Aesar Supplier No. A10889.0E
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 331753-56-1 | C8H4K2O12Sb2 | 613.83 g/mol

Antimony potassium tartrate hydrate is a compound used in the treatment of schistosomiasis. It is also used in the textile industry to aid in binding certain dyes to fabrics. It finds application in medicine as an expectorant and a nauseant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Antimony potassium tartrate hydrate is a compound used in the treatment of schistosomiasis. It is also used in the textile industry to aid in binding certain dyes to fabrics. It finds application in medicine as an expectorant and a nauseant.

Solubility
Soluble in water. Insoluble in ethanol.

Notes
Incompatible with mineral acids, strong bases, carbonates, lead, silver salts and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 331753-56-1
Molecular Formula C8H4K2O12Sb2
Molecular Weight (g/mol) 613.83
MDL Number MFCD00148863
InChI Key GUJUCWZGYWASLH-UHFFFAOYNA-J
Synonym potassium antimony tartrate hydrate, potassium antimony iii tartrate hydrate, acmc-20ajv8, antimony 3+ potassium hydrate ditartrate
IUPAC Name dipotassium 5,11-dioxo-2,6,8,12,13,14-hexaoxa-1,7-distibatricyclo[8.2.1.1⁴,⁷]tetradecane-3,9-dicarboxylate
SMILES [K+].[K+].[O-]C(=O)C1O[Sb]2OC(C(O[Sb]3OC1C(=O)O3)C([O-])=O)C(=O)O2

Specifications

Density 2.607
Assay Percent Range 98%
Quantity 2500 g
UN Number UN1551
Solubility Information Soluble in water. Insoluble in ethanol.
Formula Weight 613.82 (Anhydrous)
Percent Purity 98%
Chemical Name or Material Antimony potassium tartrate hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.