missing translation for 'onlineSavingsMsg'
Learn More

Antimony potassium tartrate trihydrate, ACS, 99.0-103.0%

Product Code. 11388568
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11388568

Brand: Thermo Scientific Alfa Aesar 040619.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 28300-74-5 | C8H4K2O12Sb2·3H2O

Antimony potassium tartrate trihydrate is used as a antiparasitic agent, which used in the treatment of schistosomiasis and leishmaniasis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28300-74-5
Molecular Formula C8H4K2O12Sb2·3H2O
MDL Number MFCD00148863
Synonym Potassium antimonyl tartrate

Specifications

Physical Form Powder
Quantity 500 g
Assay Percent Range 99.0-103.0%
Solubility Information Soluble in water and glycerine. Insoluble in ethanol.
Formula Weight 667.87 (613.82 Anhydrous)
Grade ACS
Density 2.6
Chemical Name or Material Antimony potassium tartrate trihydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.