missing translation for 'onlineSavingsMsg'
Learn More

Antimony powder, -200 mesh, 99.5% (metals basis)

Product Code. 11351138
Click to view available options
Quantity:
10 g
100 g
500 g
2 kg
Unit Size:
100g
10g
2kg
500g
This item is not returnable. View return policy

Product Code. 11351138

Brand: Thermo Scientific Alfa Aesar 010099.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7440-36-0 | Sb | 121.76 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-36-0
Molecular Formula Sb
Molecular Weight (g/mol) 121.76
MDL Number MFCD00011211 MFCD00134030
InChI Key WATWJIUSRGPENY-UHFFFAOYSA-N
Synonym powder, stibium metallicum, antimony, metallic, antimony, elemental, stibanylidyne, unii-9it35j3uv3, antymonowodor polish, unii-0vkz97k3ub, 0vkz97k3ub, antimonwasserstoffes german
PubChem CID 5354495
ChEBI CHEBI:30304
SMILES [Sb]

Specifications

Color Gray
Quantity 500 g
Mesh Size −200 Mesh
Odor Odorless
Form Powder
UN Number UN2871
IUPAC Name antimony
Percent Purity 99.5%
Assay (metals basis)
Chemical Name or Material Antimony powder
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
  • H373-May cause damage to organs.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.