missing translation for 'onlineSavingsMsg'
Learn More

Antimony(V)-chloride, 1M solution in methylene chloride, AcroSeal™, Thermo Scientific Chemicals

Product Code. 15721819 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
800mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15721819 100 mL AcroSeal™ Glass Bottle 100mL
11924201 800 mL AcroSeal™ Glass Bottle 800mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15721819 Supplier Thermo Scientific Acros Supplier No. 434521000
  • ONLINE EXCLUSIVE PRICE
    87.55 €
    81.70 € / 100mL
    Save 5.85 €
    6% Off
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7647-18-9 | Molecular Formula: Cl5Sb | Molecular Weight: 299.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Antimony(V)-chloride
CAS 7647-18-9, 75-09-2
Color Yellow
Physical Form Solution
Quantity 100 mL
Packaging AcroSeal™ Glass Bottle
Specific Gravity 1.442
Density 1.4420g/mL
Molecular Formula Cl5Sb
Linear Formula SbCl5
MDL Number MFCD00011213
Merck Index 15,686
Synonym antimony v chloride, antimony pentachloride, pentachloroantimony, antimony perchloride, antimony chloride sbcl5, antimonpentachlorid, antimoonpentachloride, perchlorure d'antimoine, pentachlorure d'antimoine, antimonpentachlorid german
InChI Key VMPVEPPRYRXYNP-UHFFFAOYSA-I
SMILES Cl[Sb](Cl)(Cl)(Cl)Cl
IUPAC Name pentachloro-$l^{5}-stibane
Molecular Weight (g/mol) 299.02
PubChem CID 24294
Formula Weight 299.01
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Antimony(V) chloride, 1M solution in methylene chloride
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H351-Suspected of causing cancer.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Dichloromethane, Antimony pentachloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.