missing translation for 'onlineSavingsMsg'
Learn More

Antipyrine, TRC

Product Code. 30137546
Change view
Click to view available options
Quantity:
50 mg
1 g
10 g
Unit Size:
10g
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137546 10 g 10g
17982615 1 g 1g
17951505 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137546 Supplier Toronto Research Chemicals Supplier No. TRCA69750010G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-80-0
Molecular Formula C11 H12 N2 O
Molecular Weight (g/mol) 188.23
Synonym Phenazone, Propyphenazone Imp. A (EP), 3H-Pyrazol-3-one, 1,2-dihydro-1,5-dimethyl-2-phenyl- (9CI, ACI), 1,2-Dihydro-1,5-dimethyl-2-phenyl-3H-pyrazol-3-one (ACI), Antipyrine (8CI), 1,5-Dimethyl-2-phenyl-1,2-dihydro-3H-pyrazol-3-one, 1,5-Dimethyl-2-phenyl-1,2-dihydropyrazol-3-one, 1,5-Dimethyl-2-phenyl-1H-pyrazol-3(2H)-one, 1,5-Dimethyl-2-phenyl-2,3-dihydro-1H-pyrazol-3-one, 1,5-Dimethyl-2-phenylpyrazol-3-one, 1-Phenyl-2,3-dimethyl-5-pyrazolone, 2,3-Dimethyl-1-phenyl-3-pyrazolin-5-one, 2,3-Dimethyl-1-phenyl-5-pyrazolone, Analgesin, Analgesine, Anodynin, Anodynine, Antipirin, Antipyrin, Apirelina, Azophen, Azophene, Dentigoa N, Dimethyloxychinizin, Dimethyloxyquinazine, Eu-Med mono, Fenazone, MeSH ID: D000983, Methozin, NSC 7945, Oxydimethylquinazine, Parodyne, Phenazon, Phenazone (pharmaceutical), Phenylon, Phenylone, Pyrazophyl, Sedatin, Sedatine
IUPAC Name 1,5-dimethyl-2-phenylpyrazol-3-one
SMILES CN1N(C(=O)C=C1C)c2ccccc2

Specifications

Quantity 10 g
InChI Formula InChI=1S/C11H12N2O/c1-9-8-11(14)13(12(9)2)10-6-4-3-5-7-10/h3-8H,1-2H3
Formula Weight 188.095
Chemical Name or Material Phenazone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.