missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Anza™ 10 DpnI

Product Code. 15595680
Change view
Click to view available options
Quantity:
4000 units
800 units
Unit Size:
1 set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15595680 800 units 1 set
15505690 4000 units 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15595680 Supplier Invitrogen™ Supplier No. IVGN0106
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Anza™ Restriction Enzyme, Anza™ 10X Buffer, Anza™ 10X Red Buffer

Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.

The Invitrogen Anza Restriction Enzyme Cloning System utilizes a single buffer system for complete digestions and incorporates Anza cloning enzymes into a clear and simple system.
• One buffer for all restriction enzymes
• One digestion protocol for all DNA types
• Complete digestion in 15 min
• Overnight digestion without star activity

Certifications

CoA

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 40 μL Anza 10 DpnI
500 μL Anza Buffer (10X)
500 μL Anza Red Buffer (10X)

Store at -20°C.
Shipping Condition Approved for shipment on Wet or Dry Ice
Methylation Sensitivity Not CpG Methylation-Sensitive
Enzyme Dpn I
Compatible Buffer Anza™ 10X Red Buffer, Anza™ 10X Buffer
Sensitive to Heat Inactivation Yes
Optimal Reaction Temperature 37°C
Type IIS RE No
Quantity 800 units
Product Type Restriction Enzyme Cloning System
Research Category Traditional Cloning
Product Line Anza
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.