missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Anza™ Alkaline Phosphatase
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
134.00 € - 452.00 €
Specifications
Shipping Condition | Approved for shipment on Wet or Dry Ice |
---|---|
Enzyme | Alkaline Phosphatase |
Compatible Buffer | 10X Buffer |
Product Type | Alkaline Phosphatase |
Product Line | Anza™ |
Includes: Anza™ Alkaline Phosphatase, Anza™ 10X Buffer, Anza™ 10X Red Buffer
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15511620
|
Invitrogen™
IVGN2208 |
2000 Reactions |
452.00 €
Each |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15545720
|
Invitrogen™
IVGN2204 |
500 Reactions |
134.00 €
1 set |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
The Invitrogen Anza Restriction Enzyme Cloning System utilizes a single buffer system for complete digestions and incorporates Anza cloning enzymes into a clear and simple system.- Utilizes the Anza 10X Buffer
- One-step digestion and dephosphorylation protocol
- Two-step dephosphorylated protocols available
- Heat inactivated in 5 minutes
Certifications
CoA
Specifications
Approved for shipment on Wet or Dry Ice | |
10X Buffer | |
Anza™ |
Alkaline Phosphatase | |
Alkaline Phosphatase |
For Research Use Only. Not for use in diagnostic procedures.