missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Anza™ T4 DNA Ligase Master Mix

Product Code. 15535720
Change view
Click to view available options
Quantity:
200 Reactions
50 Reactions
Unit Size:
1 set
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15535720 50 Reactions 1 set
15501620 200 Reactions Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15535720 Supplier Invitrogen™ Supplier No. IVGN2104
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Includes

Anza™ T4 DNA Ligase Master Mix

Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.

The Invitrogen Anza Restriction Enzyme Cloning System utilizes a single buffer system for complete digestions and incorporates Anza cloning enzymes into a clear and simple system.
• One Ligase for both cohesive and blunt end ligations
• 4X conc. enables higher volume of dilute DNA in reaction mixture
• Ligation complete in 15 minutes
• Convenient Master Mix format reduces pipetting

Certifications

CoA

TRUSTED_SUSTAINABILITY

Specifications

Concentration 4X
Content And Storage 250 μL Anza T4 DNA Ligase Master Mix

Store at -5 to -30°C.
Shipping Condition Approved for shipment on Wet or Dry Ice
Enzyme T4 DNA Ligase
Compatible Buffer Elution Buffer, 1X Anza Buffer
Quantity 50 Reactions
Product Type T4 DNA Ligase Master Mix
Product Line Anza

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.