Learn More
Thermo Scientific Chemicals Apelin-13, Human, Bovine
Description
An endogenous ligand of Apelin Receptor (APLNR/APJ receptor) that plays important roles in regulating fluid homeostasis, cardiovascular function, and insulin sensitivity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Species | Human, Bovine |
| Purification Method | HPLC |
| Content And Storage | -30°C to -10°C |
| Format | Lyophilized Powder |
| Molecular Weight (g/mol) | 1550.82 |
| Product Type | Apelin-13, human, bovine |
| Quantity | 1 mg |
| Regulatory Status | RUO - research use only |
| Sequence | Gln-Arg-Pro-Arg-Leu-Ser-His-Lys-Gly-Pro-Met-Pro-Phe |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.