missing translation for 'onlineSavingsMsg'
Learn More

Berthold Technologies Aperture 384-Well For Centro XS LB 963 Microplate Luminometer

Product Code. 31777377 Shop All Berthold Technologies Products
Change view
Click to view available options
Color:
Black
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
31777377 Black Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31777377 Supplier Berthold Technologies Supplier No. 74557
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Optional aperture to support measurement with compatible microplate formats in select microplate luminometers.

  • The Centro XS LB 963 microplate luminometer is compatible with 96-well microplates by default
  • When ordered with the 384-well aperture, it is also compatible with 384-well microplates
  • The 384-well aperture is compatible only with the Centro XS model
  • Not compatible with other Centro luminometer models

Specifications

Detection Method Luminescence
Color Black
For Use With (Application) Luminescence Applications
For Use With (Equipment) Centro XS LB 963 Microplate Luminometer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.