Learn More
Tocris Bioscience™ Aphidicolin
DNA polymerase α, δ and ε inhibitor
Brand:  Tocris Bioscience™ 5736/1
Encompass
Product Code. 15796755
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Aphidicolin is a DNA polymerase α, δ and ε inhibitor. Exhibits selectivity over DNA polymerase β and γ. Antimitotic, antibiotic and antiviral.
Specifications
Specifications
(3 R,4 R,4 aR,6 AS,8 R,9 R,11 AS,11 bS)-Tetradecahydro-3,9-dihydroxy-4,11 b-dimethyl-8,11 A-methano-11 aH-cyclohepta[a]naphthalene-4,9-dimethanol
38966-21-1