missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Aphidicolin

Product Code. 18796159
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18796159 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18796159 Supplier Tocris Bioscience™ Supplier No. 5736/1
  • ONLINE EXCLUSIVE PRICE
    174.00 €
    163.80 € / 1mg
    Save 10.20 €
    5% Off
View all promotions
 Restricted Item
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

DNA polymerase α, δ and ε inhibitor

Aphidicolin is a DNA polymerase α, δ and ε inhibitor. Exhibits selectivity over DNA polymerase β and γ. Antimitotic, antibiotic and antiviral.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (3 R,4 R,4 aR,6 AS,8 R,9 R,11 AS,11 bS)-Tetradecahydro-3,9-dihydroxy-4,11 b-dimethyl-8,11 A-methano-11 aH-cyclohepta[a]naphthalene-4,9-dimethanol
CAS 38966-21-1
Quantity 1 mg
Target RNA/DNA Polymerase Inhibitors
Molecular Formula C20H34O4
Formula Weight 338.48
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.