missing translation for 'onlineSavingsMsg'
Learn More

(+)-Aphidicolin, MP Biomedicals™

Product Code. 15997275 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15997275 1 mg 1mg
15907285 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15997275 Supplier MP Biomedicals Supplier No. 0218387301
  • ONLINE EXCLUSIVE PRICE
    168.00 €
    158.55 € / 1mg
    Save 9.45 €
    5% Off
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

By inhibiting the cellular replication of DNA polymerase II or the viral-induced DNA polymerases, Amphidocolin inhibits the growth of eukaryotic cells and certain animal viruses.

  • Aphidicolin is an antibiotic, a potent antiviral and antimitotic agent and DNA polymerase inhibitor
  • This compound is a featured product for Apoptosis research and in mitosis to research and track cellular activities

Chemical Identifiers

CAS 38966-21-1
Molecular Formula C20H34O4
Synonym aphidicolin, 38966-21-1, ICI 69653, (+)-Aphidicolin, (3 R,4 R,4 aR,6 AS,8 R,9 R,11 AS,11 bS)-4,9-bis(hydroxymethyl)-4,11 b-dimethyltetradecahydro-8,11 A-methanocyclohepta[a]naphthalene-3,9-diol

Specifications

Quantity 1 mg
Melting Point 226°C to 235°C
Color White
Purity >98
Molecular Weight (g/mol) 338.49
Chemical Name or Material (+)-Aphidicolin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.